Circulation thermostats, PRO series
Supplier: LAUDA
L000019 L000020 L000109 L000189 L000209
461-0962EA 5760 GBP
461-0962 461-0963 LAUDL000109 LAUDL000189 LAUDL000209
Circulation thermostats, PRO series
Circulating Thermostats Closed Circulator Systems Heating Circulators
The PRO heating circulation thermostats are designed for external applications up to 250 °C. The low filling volume enables rapid heating in external applications. The compact construction ensures space-saving installation of the thermostats.
  • Touchscreen can be operated with gloves
  • Low filling volume combined with Varioflex pump ensures rapid temperature changes
  • Programmer with one program, 20 segments whereas 100 programs, with 50 segments for command unit
  • Individual access rights for upto 20 users
  • Enables ramp function for easy computation