Light green SF yellowish, Sigma-Aldrich®

Supplier: SIGMA ALDRICH MICROSCOPY

Synonyms: Dihydrogen (ethyl)[4-[p-[ethyl(m-sulphonatobenzyl)amino]-p'-sulphonatobenzhydrylene]cyclohexa-2,5-dien-1-ylidene](m-sulphonatobenzyl)ammonium disodium salt, Acid Green 5, Light Green 2G, S or 2GN, CI 42095, Sodium 3-((-ethyl(-4-((4-(ethyl(3-sulphonatobenzyl)amino)phenyl)(4-sulphonatophenyl)methylene)cyclohexa-2,5-dien-1-ylidene)ammonio)methyl)benzenesulphonate, Food Green n°2, Acid Green F, Fast acid Green N, CN DcL-30

MQ200

L1886-25G
L1886-25GEA 170 GBP
L1886-25G
Light green SF yellowish, Sigma-Aldrich®
Light Green SF Yellowish
Formula: C₃₇H₃₄N₂Na₂O₉S₃
MW: 792.86 g/mol
Melting Pt: 288…289 °C
Storage Temperature: Ambient
MDL Number: MFCD00012121
CAS Number: 5141-20-8
EINECS: 225-906-5
Merck Index: 13,05506
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bf1ab1e65a89b32895082b2d8108dac47e866fb6&vid=c449be20e478fe008dd3f8a8c798a49fcf3a005a&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR