5-Fluoro-2'-deoxyuridine (floxuridine) 98

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 5-Fluoro-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 5-Fluoro-2'-deoxyuridine, Floxuridine

L16497.MCEA 50.1 GBP
L16497.MC L16497.ME
5-Fluoro-2'-deoxyuridine (floxuridine) 98
5-Fluoro-2'-deoxyuridine (Floxuridine)

Purity: 98+% CAS No.: 50-91-9 Molecular Formula: C9H11FN2O5 Formula Weight: 246.20 Melting Point: 148-151° Beilstein Reference: 2645818 Merck Reference: 14,4112 UN No.: 2811 MDL No.: MFCD00006530

Formula: C₉H₁₁FN₂O₅
MW: 246.19 g/mol
Melting Pt: 148…151 °C
Storage Temperature: Ambient
MDL Number: MFCD00006530
CAS Number: 50-91-9
EINECS: 200-072-5
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR