Methyl acetate 99

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Acetic acid methyl ester

L14475.AP L14475.AU L14475.0F
L14475.APEA 23.9 GBP
L14475.AP L14475.AU L14475.0F
Methyl acetate 99
Methyl acetate

Purity: 99% CAS No.: 79-20-9 Molecular Formula: CH3COOCH3 Formula Weight: 74.08 Flash Point: -10°(14°F) Refractive Index: 1.3620 Odour: Pleasant Melting Point: -98° Boiling Point: 56-58° Beilstein Reference: 1736662 Merck Reference: 14,6008 UN No.: 1231 MDL No.: MFCD00008711

Formula: C₃H₆O₂
MW: 74.08 g/mol
Boiling Pt: 56…58 °C
Melting Pt: –98 °C
Density: 0.934
Flash Pt: –10 °C (14 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008711
CAS Number: 79-20-9
EINECS: 201-185-2
UN: 1231
ADR: 3 II
Merck Index: 13,06038

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR