Sodium mercaptoacetate (sodium thioglycolate) 97

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Thioglycolic Acid, Thioglycolic Acid Sodium Salt, Mercaptoacetic acid, sodium salt

L14356.14 L14356.22 L14356.36
L14356.14EA 25.7 GBP
L14356.14 L14356.22 L14356.36
Sodium mercaptoacetate (sodium thioglycolate) 97
Sodium mercaptoacetate (Sodium thioglycolate)

Purity: 97% CAS No.: 367-51-1 Molecular Formula: HSCH2CO2Na Formula Weight: 114.10 Odour: Stench pH: ~6 Melting Point: ca 305° Beilstein Reference: 4569109 Merck Reference: 14,8692 UN No.: 3335 MDL No.: MFCD00043386

Formula: HSCH₂COONa
MW: 114.1 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00043386
CAS Number: 367-51-1
EINECS: 206-696-4
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR