(S)-(+)-β-Aminophenethyl alcohol 98

Supplier: Thermo Fisher Scientific
Warning

Synonyms: (S)-(+)-2-Phenylglycinol, L-Phenylglycinol, (S)-2-Amino-2-phenylethanol

L13265.03 L13265.06 L13265.14
L13265.03EA 30.6 GBP
L13265.03 L13265.06 L13265.14
(S)-(+)-β-Aminophenethyl alcohol 98
(S)-(+)-β-Aminophenethyl alcohol

Purity: 98+% CAS No.: 20989-17-7 Molecular Formula: C8H11NO Formula Weight: 137.18 Melting Point: 76-78° Beilstein Reference: 3196190

Formula: C₈H₁₁NO
MW: 137.18 g/mol
Melting Pt: 76…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00064404
CAS Number: 20989-17-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR