Hydrochloric acid 36% w/w aqueous solution

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Hydrochloric acid solution, Muriatic acid

L13091.AU L13091.0F
L13091.AUEA 29.3 GBP
L13091.AU L13091.0F
Hydrochloric acid 36% w/w aqueous solution
Hydrochloric acid

CAS No.: 7647-01-0 Molecular Formula: HCl Formula Weight: 36.46 Odour: Pungent Boiling Point: 81.5-110° Merck Reference: 14,4780 MDL No.: MFCD00011324

Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 81.5…110 °C
Melting Pt: –30 °C
Density: 1.19
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8 II
Merck Index: 14,4780

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR