Methyl oleate, technical C18 71-90%, C18:1 >65% of C18

Supplier: Thermo Fisher Scientific

Synonyms: cis-Oleic Acid Methyl Ester, Oleic acid methyl ester

L02937.AK L02937.AU
L02937.AKEA 20.5 GBP
L02937.AK L02937.AU
Methyl oleate, technical C18 71-90%, C18:1 >65% of C18
Methyl oleate

CAS No.: 112-62-9 Molecular Formula: C19H36O2 Formula Weight: 296.50 Flash Point: 163°(325°F) Refractive Index: 1.4520 Odour: Weak, characteristic Boiling Point: 168-170°/2mm Beilstein Reference: 1727037 Merck Reference: 14,6828 MDL No.: MFCD00009578

Formula: C₁₉H₃₆O₂
MW: 296.49 g/mol
Boiling Pt: 168…170 °C (2 mmHg)
Melting Pt: –20 °C
Density: 0.876
Flash Pt: 163 °C (325 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009578
CAS Number: 112-62-9
EINECS: 203-992-5
Merck Index: 14,6828

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR