Platinum 10% on carbon, dry

Supplier: Thermo Fisher Scientific
Warning

Synonyms: Platinium

L00469.03 L00469.06 L00469.14
L00469.03EA 40.4 GBP
L00469.03 L00469.06 L00469.14
Platinum 10% on carbon, dry
Platinum

CAS No.: 7440-06-4 Odour: Odourless UN No.: UN1325 MDL No.: MFCD00011179

Formula: Pt
MW: 195.08 g/mol
Boiling Pt: 3827
Melting Pt: 1769
Storage Temperature: Ambient
MDL Number: MFCD00011179
CAS Number: 7440-06-4
EINECS: 231-116-1
UN: 1325
ADR: 4.1 III
Merck Index: 14,7529

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR