3,5-Di-tert-butyl-o-benzoquinone 98

Supplier: Thermo Fisher Scientific
Warning

L00169.06 L00169.14
L00169.06EA 60.9 GBP
L00169.06 L00169.14
3,5-Di-tert-butyl-o-benzoquinone 98
3,5-Di-tert-butyl-o-benzoquinone

Purity: 98+% CAS No.: 3383-21-9 Molecular Formula: C14H20O2 Formula Weight: 220.31 Melting Point: 113-117° Beilstein Reference: 2047944

Formula: C₁₄H₂₀O₂
MW: 220.31 g/mol
Melting Pt: 113…117 °C
Storage Temperature: Ambient
MDL Number: MFCD00001647
CAS Number: 3383-21-9
EINECS: 222-189-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR