Reaction vessels, cylindrical, with flat bottom and thermal jacket
Supplier: KGW
TRGN7399
KGWATRGN7399EA 0 GBP
KGWATRGN7399
Reaction vessels, cylindrical, with flat bottom and thermal jacket
Reaction Vessels Reaction Vessels, Cylindrical Shape
This cylindrical reaction vessel is made up of borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
It has flat bottom and thermal jacket, GL 18 Olive tubing connection.
Order Now
SPECIFICATIONS
Environmentally Preferable