Ear defenders, Sonis®
Supplier: JSP
AEB030-0CY-000 AEB010-0AY-800 AEB020-0AY-900 AEB030-0AY-000 AEB040-0A1-A00
JSPAAEB030-0CY-000EA 0 GBP
JSPAAEB030-0CY-000 JSPAAEB010-0AY-800 JSPAAEB020-0AY-900 JSPAAEB030-0AY-000 JSPAAEB040-0A1-A00
Ear defenders, Sonis®
Earmuffs Noise Blocking Earmuffs
The Sonis® ear defender range has been designed with a colour coded system for simple selection, and peaks at an unbeatable 37 SNR.
  • Foam/PET combination cushions
  • ABS cups
  • Optimum cushion pressure and comfort
  • Adjustable steel arms
  • Hygienic
The Sonis® range of ear defenders have been designed by the JSP R&D team. Working with a leading UK acoustic engineering research facility they have developed an ear defender range that peaks at an unbeaten 37 SNR.
The over-moulded headband has a built-in comfort area with ventilation. The headband force andMore Product Information