Sodium dodecyl sulphate (SDS) 97% for electrophoresis

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Dodecyl sulphate sodium salt, Sodium dodecyl sulfate, Dodecyl sulphuric acid sodium salt, Lauryl sulphate sodium salt, Sodium lauryl sulphate

J64241.14 J64241.22 J64241.36
J64241.14EA 33.2 GBP
J64241.14 J64241.22 J64241.36
Sodium dodecyl sulphate (SDS) 97% for electrophoresis
Sodium dodecyl sulphate (SDS)

Purity: 98%
CAS No.: 151-21-3
Molecular Formula: C12H25NaO4S
Formula Weight: 288.38
Physical Form: Powder
Appearance: White
Melting Point: ca 206°
Solubility: Soluble in water
Beilstein Reference: 3599286
Merck Reference: 14,8636
UN No.: 2926
MDL No.: MFCD00036175

Formula: CH₃(CH₂)₁₁OSO₃Na
MW: 288.38 g/mol
Melting Pt: ∼206 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
EINECS: 205-788-1
UN: 1325
ADR: 4.1 III
Merck Index: 14,8636
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe4cb1741fba41f699a3be006d0b01b1b551898c&vid=c6df9a86fc351b319e68cc79c1c17f733b4fec67&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR