N(α)-Acetyl-L-tryptophan 99

Supplier: Thermo Fisher Scientific

Synonyms: N-Acetyl-L-tryptophan, Ac-Trp-OH

J64096.06 J64096.14
J64096.06EA 30.9 GBP
J64096.06 J64096.14
N(α)-Acetyl-L-tryptophan 99
N(α)-Acetyl-L-tryptophan
Formula: C₁₃H₁₄N₂O₃
MW: 246.3 g/mol
Melting Pt: 186 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00065976
CAS Number: 1218-34-4
EINECS: 214-935-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR