L-Threonine Cell Culture

Supplier: Thermo Fisher Scientific

Synonyms: (2S,3R)-(-)-Threonine, H-Thr-OH, (2S,3R)-2-Amino-3-hydroxybutyric acid

J63709.30 J63709.A1
J63709.30EA 60.4 GBP
J63709.30 J63709.A1
L-Threonine Cell Culture
L-Threonine

CAS No.: 72-19-5 Molecular Formula: C4H9NO3 Formula Weight: 119.12 Physical Form: Powder Appearance: White Melting Point: 256° dec. Beilstein Reference: 1721646 Merck Reference: 14,9380 MDL No.: MFCD00064270

Formula: C₄H₉NO₃
MW: 119.12 g/mol
Melting Pt: 256 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00064270
CAS Number: 72-19-5
EINECS: 200-774-1
Merck Index: 14,9380

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR