Alpha Sterile Kits
Supplier: ISOFIELD IRELAND
80551S 80551M 80551L 80551XL 80551XXL 80551XXXL
ISOF80551SEA 1080 GBP
ISOF80551S ISOF80551M ISOF80551L ISOF80551XL ISOF80551XXL ISOF80551XXXL
Alpha Sterile Kits
Disposable Garments
Sterile coverall with mandarin collar, hood, and overboots.
  • Manufactured in a facility operating under ISO 9001:2015 quality management system
  • Processed in a NEBB certified ISO Class 5 cleanroom
Mandarin collar coverall:
GMP Grade A and B, ISO Class 4 and 5 cleanroom compatible
Two-layer mandarin collar provides secure fit at neck area
Zip flap with double-sided tape prevents ingress and egress of particles
Low-linting, anti-static fabric suitable for chemo handling
Elasticated thumb loops
Back elastic to prevent bellows effect
Hood:
Latex-free enclosedMore Product Information