Rabbit IgG Isotype Control (RPE (R-Phycoerythrin))

Supplier: ImmunoReagents
IR2800
IMMRIR2800EA 494 GBP
IMMRIR2800
Rabbit IgG Isotype Control (RPE (R-Phycoerythrin))
Antibodies
R-phycoerythrin is used in immunoassays such as FACS, flow cytometry, multimer/tetramer applications. With new instrumetation available, R-PE is also well suited for immunohistochemistry (IHC) and Western blot applications.

Type: Control
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Goat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR