Anti-IgG Mouse Polyclonal Antibody (Unconjugated)

Supplier: ImmunoReagents
IR2374
IMMRIR2374EA 216 GBP
IMMRIR2374
Anti-IgG Mouse Polyclonal Antibody (Unconjugated)
Antibodies
Type:
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR