Anti-IgG Goat Polyclonal Antibody

Supplier: ImmunoReagents
IR2136
IMMRIR2136EA 91.7 GBP
IMMRIR2136
Anti-IgG Goat Polyclonal Antibody
Antibodies
ImmunoReagents is a manufacturer of quality research and production reagents for the Life Science Industries. Product lines include a wide range of antibodies used in ELISA, immunohistochemistry (IHC), western blotting, flow cytometry (FACS), immunofluorescence and microscopy.  Antibodies produced cover a broad range of Immunoglobulins from various species cross absorbed to provide high specificity.  

  • Origin: US

Products will be accompanied with a standard product CoA to include application, purity and cross reactivityMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol IgG
Antigen name Immunoglobulin G
Environmentally Preferable
Antibody type Secondary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Rabbit
Host Goat
Isotype IgG
Western blot Yes
ELISA Yes
ImmunoChemistry Yes
ImmunoPrecipitation Yes
ImmunoFluorescence Yes
Flow cytometry Yes
Immunogen Rabbit IgG
Format Purified IgG
Form Clear, colourless liquid, 0,2 µm filtered
Purification Affinity purified using solid phaseRabbit IgG
Concentration >4,5 mg/ml (E 1% at 280 nm = 14,0)
Cross adsorption Yes
Storage buffer 10 mM sodium phosphate, 0,15 M sodium chloride, pH 7,2
Storage temperature 2...8 °C
Shipping temperature 2...8 °C
Tested applications Fluorescence-Activated Cell Sorting, Immunodiffusion, Labeling

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR