Anti-IgG Hamster Polyclonal Antibody

Supplier: ImmunoReagents
HA-003-ZBIO
IMMRHA-003-ZBIOEA 101 GBP
IMMRHA-003-ZBIO
Anti-IgG Hamster Polyclonal Antibody
Antibodies
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Hamster
Isotype:
Reactivity:
Order Now

SPECIFICATIONS

Antigen symbol IgG
Environmentally Preferable
Antibody type Secondary
Clonality Polyclonal
Reactivity
Host Hamster
Isotype
Form Liquid
Concentration 1.5 mg/ml
Shipping temperature 2-8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR