CBC ROTAVISC lo-vi Viscometer Package
Supplier: IKA
10014047
IKAA10014047EA 0 GBP
IKAA10014047
CBC ROTAVISC lo-vi Viscometer Package
Viscometers Rotational Viscometers
The viscosity of most liquids depends on the temperature. To obtain reliable measurement results, accurate temperature control is crucial. The CBC ROTAVISC lo-vi Package is the perfect solution for precise viscosity measurement of temperature-dependent samples in the laboratory.
  • Save time: A second beaker (600 ml) can be pre-tempered during the measurement
  • Perfect results: Optimised water flow management in the insulated bath for fast and precise temperature control
  • Improved ecological footprint: Equipped with the environmentally friendly natural
More Product Information