Handheld digital microscopes, Dino-Lite High Magnification
Supplier: Dino-Lite / IDCP B.V.
AM4013MZT4 AM4113ZT4 AM4013MT5 AM4113T5 AM4515T5 AM4515T8 AM4515ZT4 AM7515MT4A AM7515MT8A AM73515MT8A
IDCPAM4013MZT4EA 659 GBP
IDCPAM4013MZT4 IDCPAM4113ZT4 IDCPAM4013MT5 IDCPAM4113T5 IDCPAM4515T5 IDCPAM4515T8 630-2750 IDCPAM7515MT4A IDCPAM7515MT8A IDCPAM73515MT8A
Handheld digital microscopes, Dino-Lite High Magnification
Microscopes
Dino-Lite models within the High Magnification series exceed a magnification of 200 times and offer magnifications of 400×, 500× or even 900 times. Magnification range: 400 - 900×.
  • High speed USB 2.0 connection
  • Metal housing or composite housing
  • Edge series models with high-quality optics provide a very sharp, bright and natural colour image with very low aberration and vignetting
  • Wireless streaming to a tablet, smartphone, or computer is possible in combination with WF-10 unit (to be ordered separately)
  • Integrated polariser greatly reduces glare on
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable