Serum-free medium, MP-MRC-5®, MP Biomedicals
Supplier: MP Biomedicals
092013426
ICNA092013426EA 1210 GBP
ICNA092013426
Serum-free medium, MP-MRC-5®, MP Biomedicals
Cell Culture Media
MP-MRC-5® medium was developed specifically for the culture of MRC-5 cells and microcarrier culture of MRC-5 cells in production systems.
  • Serum-free, animal component free
  • Very low protein (<90 ng/ml)
  • Specific for MRC-5 cells
Order Now
SPECIFICATIONS
Environmentally Preferable