Ampicillin trihydrate, Crystalline

Supplier: MP Biomedicals
Danger

Synonyms: (2S,5R,6R)-6-((R)-2-Amino-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid trihydrate

0219014725
ICNA0219014725EA 132 GBP
ICNA0219014725
Ampicillin trihydrate, Crystalline
Ampicillin trihydrate

  • At 25 °C ,pKa = 2.5 (-COOH),pKa = 7.3 (-NH2) (Lit.)


Storage temperature.: +4 °C

Formula: C₁₆H₁₉N₃O₄S·3H₂O
MW: 403.45 g/mol
MDL Number: MFCD00072036
CAS Number: 7177-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR