Strontium chloride hexahydrate, powder_crystalline ACS

Supplier: MP Biomedicals

0215258390
ICNA0215258390EA 115 GBP
ICNA0215258390
Strontium chloride hexahydrate, powder_crystalline ACS
Strontium chloride hexahydrate
Formula: SrCl₂·6H₂O
MW: 266.62 g/mol
Boiling Pt: 100
Melting Pt: 61
MDL Number: MFCD00149865
CAS Number: 10025-70-4
Merck Index: 13,08920

Order Now

Specification Test Results

Synonyms Strontium Dichloride Hexahydrate
Form White, Hard Packed, Crystalline Powder
Storage Store at Room Temperature (15 - 30 °C)
Assay 99,0 - 103,0%
Heavy Metals NMT 0,0005% ; Iron NMT 0,0005%
Insoluble Matter NMT 0,005%
pH 5,0 - 7,0 (5% sol)
Sulfate NMT 0,001%
Barium NMT 0,05%
calcium NMT 0,05%
Magnesium NMT 0,0002%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR