Serving Trolley Ergo
Supplier: HUPFER METALLWERKE
0133713
HUPF0133713EA 0 GBP
HUPF0133713
Serving Trolley Ergo
Trolleys
Trolley in stable, self-supporting and hygienic construction made of high-quality stainless steel. Round tubular frame with firmly welded, deep-drawn borders with profile edges, sound-absorbing lining, hygienically folded and folded inwards, in an easy-to-clean design. Top shelf as standard with reinforcing beam, consisting of two longitudinal and one diagonal strut, which are fully welded together.
  • With Ergo - Handle
  • Payload: 140 kg
  • Shelf clearance: 278 mm
  • Stainless steel
Push bars integrated into the tubular frame on both sides ensure easyMore Product Information