Single-use syringes, 3-piece, HENKE-JECT®
Supplier: HENKE SASS WOLF
5010.200V0 5010.A00V0 5100.000V0 8300005761 8300005762 5050.000V0 5050.X00V0 5100.X00V0 5200.000V0 5200.X00V0 8300006682 8300006681 8300006680 5202D000V0 8300005458 5100.090D0 5100.X90D0 8300014579 8300019869 8300005157 5300.000D0 5300.X00D0 8300063477 8300065154 8300063469 8300063470 8300063472 8300065151 8300065156 8300065153 8300065157 8300065152 8300063471 8300063465
613-2040EA 23.3 GBP
613-2040 613-2041 613-2044 613-2050 613-2051 613-2042 613-2043 613-2045 613-2046 613-2047 613-2053 613-2052 613-1586 HSWA5202D000V0 615-0042 HSWA5100.090D0 613-6080 613-2794 613-3135 613-6079 613-6913 613-6914 HSWA8300063477 HSWA8300065154 HSWA8300063469 HSWA8300063470 HSWA8300063472 HSWA8300065151 HSWA8300065156 HSWA8300065153 HSWA8300065157 HSWA8300065152 HSWA8300063471 HSWA8300063465
Single-use syringes, 3-piece, HENKE-JECT®
Syringes Syringes, Medical
Disposable, three-piece syringes with PP barrel and piston, polyisoprene stopper, latex-free.
  • Lubricated synthetic rubber piston allows a smooth draw
  • Available in non sterile version (bulk) upon request
Without needles: Luer lock types are compatible for use with most common syringe pumps. Please ask for further information.
With fixed needles: Needle tube permanently attached in syringe, no dead space, safety stop, suitable for dosage of very small volumes.
Luer lock types are compatible for use with most common syringe pumps. Please ask forMore Product Information