Surgical masks, type IIR
Supplier: Medicom
M36101-30
HOPEM36101-30EA 240 GBP
HOPEM36101-30
Surgical masks, type IIR
Masks Surgical Masks
Non-woven multi-layer construction offers excellent resistance to human liquid/fluid projection at a pressure of 120 m mHg.
  • Bacterial filtration efficiency is greater than 98%
  • Two comfortable large ear loops
  • Very soft polypropylene non-woven edge
  • Anatomic folding limits inward leakage
  • Integrated anti-fog nose piece
  • Filter without micro glass
  • Manufactured in ISO 8 cleanroom (particle and microbiological control)
  • Made in France
Designed for use by operating room personnel for surgeries with a risk of biological fluid projection. The single-use, nonMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable