Medical Masks, Type II Tie-On
Supplier: Medicom
M24101-30
HOPEM24101-30EA 69.1 GBP
HOPEM24101-30
Medical Masks, Type II Tie-On
Masks Surgical Masks
Medical Device: Class I - Medical mask type: Type II Tie-On
  • The Kolmi® Feeldry range features an ultra-absorbent inner layer, ensuring optimal and consistent comfort, even during prolonged use
  • Tested according to BFE, VFE, and PFE standards
Tested according to BFE, VFE, and PFE standards.
Certifications:
EN14683:2019+AC:2019 standard