Overshoes Non woven, PP
Supplier: Medicom
H10020
HOPEH10020EA 201 GBP
HOPEH10020
Overshoes Non woven, PP
Disposable Garments
These non woven overshoes are made up of PP with Herringbone print sole.
  • Its polypropylene material is particularly used on smooth surfaces in dry environments
  • Its chevron patterns ensure grip on dry floors
Certifications: ISO 9001 manufacturing site.
Order Now
SPECIFICATIONS
Environmentally Preferable
Gender
Garment application
Weight 40 g/m²