Pharmaceutical analytical standard, Colterol-(tert-butyl-d9) acetate, reference material, Honeywell Fluka™

Supplier: Honeywell Chemicals

R1210-10MG
HONCR1210-10MGEA 525 GBP
HONCR1210-10MG
Pharmaceutical analytical standard, Colterol-(tert-butyl-d9) acetate, reference material, Honeywell Fluka™
Standards Organic Standards

High quality inorganic and organic reference materials produced according to ISO 9001.


  • Ensure you have the information you need to quickly start your analysis, and pass relevant audits
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results

Order Now

SPECIFICATIONS

Description Colterol-(tert-butyl-d₉) acetate
Environmentally Preferable
CAS No. 1420043-63-5
Formula C₁₂D₉H₁₀NO₃ · C₂H₄O₂
Melting point 157...161 °C
Freezing point 158 - 160 °C
Storage temperature 2...8 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR