Cadmium ICP standards, (CRMs), Fluka™
Supplier: Honeywell Chemicals
E1139-100ML
HONCE1139-100MLEA 114 GBP
HONCE1139-100ML
Cadmium ICP standards, (CRMs), Fluka™
Standards Cationic Standards Cadmium Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certifications: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).