Methanol Reag. Ph. Eur., ACS, ISO, Burdick & Jackson™

Supplier: Honeywell Chemicals
Danger

Synonyms: Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol

65757-190L
HONC65757-190LEA 707 GBP
HONC65757-190L
Methanol Reag. Ph. Eur., ACS, ISO, Burdick & Jackson™
Methanol
Formula: CH₃OH
MW: 32.04 g/mol
Boiling Pt: 64…65 °C
Melting Pt: -98
Density: 0.79 g/cm3
Flash Pt: 11 °C
MDL Number: MFCD00004595
CAS Number: 67-56-1
UN: 1230
ADR: 3 II
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.8 %
Boiling range 64 - 65 °C
Density (D 20/20) 0.791 - 0.793
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.05 %
Free acid (as HCOOH) Max. 0.002 %
Free alkali (as NH₃) Max. 0.00005 %
Aluminium (Al) Max. 0.00005 %
Boron (B) Max. 0.000002 %
Barium (Ba) Max. 0.00001 %
Bismuth (Bi) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000005 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000001 %
Iron (Fe) Max. 0.00001 %
Lithium (Li) Max. 0.00001 %
Magnesium (Mg) Max. 0.00001 %
Manganese (Mn) Max. 0.000001 %
Molybdenum (Mo) Max. 0.00001 %
Sodium (Na) Max. 0.00005 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.000002 %
Tin (Sn) Max. 0.00001 %
Strontium (Sr) Max. 0.00001 %
Zinc (Zn) Max. 0.00001 %
Chloride (Cl) Max. 0.00005 %
Sulfate (SO₄) Max. 0.0001 %
KMnO₄ reducing matter (as O) Max. 0.00025 %
Acetone (GC) Max. 0.001 %
Acetaldehyde Max. 0.001 %
Benzene (GC) Max. 0.0002 %
Ethanol (GC) Max. 0.1 %
Formaldehyde Max. 0.0001 %
Carbonyl compounds (as CO) Max. 0.005 %
Mixable with H₂O complying
Reaction against H₂SO₄ complying
APHA Max. 10
Appearance complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR