Nitric acid ≥65%, Puriss. p.a. Reag. Ph. Eur., ISO Determination_with dithizone, Fluka™

Supplier: Honeywell Chemicals
Danger

Synonyms: Nitric acid solution

30709-2.5L-GL 30709-1L 30709-1L-GL 30709-2.5L 30709-4X2.5L 30709-4X2.5LGL 30709-4X2.5L-G 30709-190L 30709-35KG 30709-990L
30709-2.5L-GLEA 95.9 GBP
30709-2.5L-GL 30709-1L 30709-1L-GL 30709-2.5L 30709-4X2.5L 30709-4X2.5LGL 30709-4X2.5L-GL HONC30709-190L HONC30709-35KG HONC30709-990L
Nitric acid ≥65%, Puriss. p.a. Reag. Ph. Eur., ISO Determination_with dithizone, Fluka™
Nitric acid
Formula: HNO₃
MW: 63.01 g/mol
Boiling Pt: 122 °C (1.013 hPa)
Melting Pt: –31 °C
Density: 1.400…1.480 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 II
Merck Index: 13,06608

Order Now

Specification Test Results

Appearance complying
Assay 65.0 - 67.0 %
Density (D 20/4) 1.390 - 1.410
Residue on ignition (SO₄) Max. 0.0005 %
Silver (Ag) Max. 0.000001 %
Arsenic (As) Max. 0.000001 %
Aluminium (Al) Max. 0.000005 %
Barium (Ba) Max. 0.000001 %
Beryllium (Be) Max. 0.000001 %
Bismuth (Bi) Max. 0.00001 %
Calcium (Ca) Max. 0.00001 %
Cadmium (Cd) Max. 0.000001 %
Cobalt (Co) Max. 0.000001 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000001 %
Iron (Fe) Max. 0.00001 %
Germanium (Ge) Max. 0.000005 %
Mercury (Hg) Max. 0.000001 %
Potassium (K) Max. 0.000005 %
Lithium (Li) Max. 0.000001 %
Magnesium (Mg) Max. 0.00001 %
Manganese (Mn) Max. 0.000001 %
Molybdenum (Mo) Max. 0.000002 %
Sodium (Na) Max. 0.00005 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.000001 %
Strontium (Sr) Max. 0.000001 %
Titanium (Ti) Max. 0.00001 %
Thallium (Tl) Max. 0.000005 %
Vanadium (V) Max. 0.000001 %
Zinc (Zn) Max. 0.000005 %
Zirconium (Zr) Max. 0.00001 %
Heavy metals (as Pb) Max. 0.0002 %
Chloride (Cl) Max. 0.00005 %
Phosphate (PO₄) Max. 0.00005 %
Sulfate (SO₄) Max. 0.00005 %
Suitability for determinations with dithizone complying
Residual solvents complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR