Automatic burettes, AS, Pellet pattern, lateral stopcocks and intermediate valve
Supplier: HIRSCHMANN
355036060 355037060 355037560
HIRS355036060EA 456 GBP
HIRS355036060 HIRS355037060 HIRS355037560
Automatic burettes, AS, Pellet pattern, lateral stopcocks and intermediate valve
Burettes
Automatic burettes, Pellet pattern, class AS, brown glass, DURAN® with white graduation.
  • Main point ring graduation
  • Lateral PTFE stopcock with intermediate valve
Certifications: Compliance with DIN EN ISO 385.