Avantor® Hichrom PEEKsil™, PEEK Tubing
Supplier: Avantor
HICHHI-260EA 98.3 GBP
HICHHI-260 HICHHI-261 HICHHI-382
Avantor® Hichrom PEEKsil™, PEEK Tubing
Tubing Chromatography Tubing
Precision bore PEEK tubing is designed specifically for modern LC, LCMS and automation usage.
  • Chemical resistant and inert to most commonly used solvents
PEEK tubing is a flexible alternative to stainless steel tubing in high pressure applications. Also ensures reproducibility in chromatographic flow rates through smooth internal surface along with consistent and tight toleranced IDs.
Order Now
SPECIFICATIONS
Environmentally Preferable