Avantor® Hichrom, Tubing Cutters, Stainless Steel
Supplier: Avantor
HICHHI-192EA 281 GBP
HICHHI-192
Avantor® Hichrom, Tubing Cutters, Stainless Steel
Cutters
Stainless steel cutters are specifically designed for ¹/₁₆" stainless steel tubing.
  • Smooth and uniform cuts