PEEKsil™, Chromatography Tubing, Precut
Supplier: Avantor
HICH05-SS-05EA 46 GBP
HICH05-SS-05 HICH05-SS-10 HICH10-SS-07 HICH10-SS-10 HICH10-SS-30 HICH20-SS-10
PEEKsil™, Chromatography Tubing, Precut
Tubing Chromatography Tubing
316 Stainless steel, precision bore ¹/₁₆" type tubing.
  • Passivated for chemical resistance
  • Square, burr-free ends
  • Precut lengths
0,005 and 0,007" Int.Ø- suitable for minimal dead volume injector-column or column-detector connections of microbore or short columns containing 3 µm material. Sample and material must be filtered to avoid blockage.
0,010" Int.Ø - suitable for connection of analytical and semi-preparative columns containing 5 µm and 10 µm material. It should be used in all parts of the system where dead volume needs to be kept to minimum.
More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable