ICP-MS Nebulisers and Supplies for PerkinElmer
Supplier: Agilent
8003-0475 8003-0476 8003-0477 8003-0478 8003-0479 8003-0480 8003-0481 8003-0483 8003-0484 8003-0485 8003-0486 8003-0487 8003-0488 8003-0489 8003-0490 8003-0492 8003-0493 8003-0495 8003-0500 8003-0502 8003-0503 8003-0504 8003-0505 8003-0506 8003-0507 8003-0508 8003-0511 8003-0512 8003-0513 8003-0589
HEWL8003-0475EA 0 GBP
HEWL8003-0475 HEWL8003-0476 HEWL8003-0477 HEWL8003-0478 HEWL8003-0479 HEWL8003-0480 HEWL8003-0481 HEWL8003-0483 HEWL8003-0484 HEWL8003-0485 HEWL8003-0486 HEWL8003-0487 HEWL8003-0488 HEWL8003-0489 HEWL8003-0490 HEWL8003-0492 HEWL8003-0493 HEWL8003-0495 HEWL8003-0500 HEWL8003-0502 HEWL8003-0503 HEWL8003-0504 HEWL8003-0505 HEWL8003-0506 HEWL8003-0507 HEWL8003-0508 HEWL8003-0511 HEWL8003-0512 HEWL8003-0513 HEWL8003-0589
ICP-MS Nebulisers and Supplies for PerkinElmer
Chromatography Supplies Chromatography Spare Parts
Key design features of nebulisers for PerkinElmer ICP-MS systems provide the best precision, efficient drainage, minimal burner blockage, and reduced interferences.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the stand behind warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
  • Many parts in packaging designed to be more durable, sturdy and provide the best protection during shipping to ensure the part arrives with no damage
The GemClean Cross-Flow IIMore Product Information