Amino acids analysis standards, AdvanceBio AAA
Supplier: Agilent
5061-3330 5061-3331 5061-3332 5061-3333 5061-3334 5061-3335 5061-3337 5061-3339 5062-2478 5062-2479 5190-9426
HEWL5061-3330EA 0 GBP
HEWL5061-3330 HEWL5061-3331 HEWL5061-3332 HEWL5061-3333 HEWL5061-3334 HEWL5061-3335 HEWL5061-3337 HEWL5061-3339 HEWL5062-2478 HEWL5062-2479 HEWL5190-9426
Amino acids analysis standards, AdvanceBio AAA
Standards Biological Standards
A kit with amino acid standards (17 components) at different concentrations together with reagents and buffers required for analysis.