High performance, manual defrost, −20 °C freezers, TSX

Supplier: Thermo Scientific
TSX2320FV TSX2320EV TSX3020FV
HERATSX2320FVEA 7010 GBP
HERATSX2320FV HERATSX2320EV HERATSX3020FV
High performance, manual defrost, −20 °C freezers, TSX
Freezers Laboratory / General Purpose Freezers
The manual defrost, high-performance freezers are designed for applications in which even slight intermittent coil warming during auto defrost cannot be tolerated. The Series’ V-drive technology is designed to provide temperature uniformity that continually adapts to user patterns, offering significant energy savings without compromising protection.

  • Cold wall convection cooling with temperature uniformity
  • Enzyme freezer features enzyme bins
  • Quiet operation at just 50 dBA
  • Keyed on/off switch with easy to use, setpoint security
  • Self-closing door with 90°
More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR