Rotavac Vacuum Pumps
Supplier: Heidolph Instruments GmbH & Co.KG
591-00160-00 591-00141-00 591-00141-03 591-00141-04 591-00142-00 591-00160-03
531-0457EA 2300 GBP
531-0457 HEID591-00141-00 HEID591-00141-03 HEID591-00141-04 HEID591-00142-00 HEID591-00160-03
Rotavac Vacuum Pumps
Pumps Vacuum Rotary Evaporator Pumps
Rotavac Vario
Three stage diaphragm pump is made from chemically resistant material.
For all Hei-VAP models
Precise vacuum control avoids bumping and eliminates foaming of your evaporation solution
Due to automatic vacuum supply to the process parameters you have more time for important laboratory activities
Can be combined with a condenser
Rotavac Valve Tec
Two-stage diaphragm pump is made from chemically resistant material.
For all Hei-VAP models
Recommended for solvents with low or medium boiling points
Vacuum can be controlled manually or via valve operated vacuum controllers
Can be combinedMore Product Information