Centrifuge tubes with screw cap, round bottom, AR-Glas®, Assistent®
Supplier: GLASWARENFABRIK KARL HECHT
40976010
HECH40976010EA 142 GBP
HECH40976010
Centrifuge tubes with screw cap, round bottom, AR-Glas®, Assistent®
Tubes Centrifuge Tubes
Cylindrical, ungraduated, with thread GL 14.
  • Round bottom
The screw caps are gas-sterilisable up to +121 °C using ethylene oxide.
Order Now
SPECIFICATIONS
Environmentally Preferable