Waste bins, WIVA™ VAT-5
Supplier: HANNELLS INDUSTRIER
030V5090BL 050V5090BL
HANL030V5090BLEA 0 GBP
HANL030V5090BL HANL050V5090BL
Waste bins, WIVA™ VAT-5
Bins Waste Bins
WIVA™ VAT-5 medical waste containers provide safe storage and transport of healthcare hazardous waste.
  • Fit for use in various medical environments, also for hygienic autoclaving and incineration processes
  • Simple but robust closing system
Certifications: UN approved for Class 6.2 infectious waste, UN 3291
Order Now
SPECIFICATIONS
Environmentally Preferable