Analogue stopwatches, split-second addition timer
Supplier: HANHART
135.0201-0S 135.0301-0S 135.0101-0S 135.0401-0S
HANH135.0201-0SEA 754 GBP
HANH135.0201-0S 609-0146 609-1129 609-1132
Analogue stopwatches, split-second addition timer
Timers Analogue Stopwatches
Addition timer for time measurement with interrupt option, also drag indicator for split-time measurement.
  • Chrome plated brass casing, diamond cut
  • Lever, 7 jewels
  • Shock resistant, additional dust and water protection
Order Now
SPECIFICATIONS
Environmentally Preferable