Baths and circulators, stainless steel, SAHARA series
Supplier: THERMO ELECTRON LED
1541131 152-1491 152-1301 152-1211 153-1211 153-1131 153-1071 153-1031 1541071 1541491 156-1071 1561451 1551451 1521451 1531451
HAKA1541131EA 3720 GBP
HAKA1541131 HAKA152-1491 HAKA152-1301 HAKA152-1211 HAKA153-1211 HAKA153-1131 HAKA153-1071 HAKA153-1031 HAKA1541071 HAKA1541491 HAKA156-1071 HAKA1561451 HAKA1551451 HAKA1521451 HAKA1531451
Baths and circulators, stainless steel, SAHARA series
Circulating Thermostats Open Circulator Systems Baths with Detachable Circulators
The SAHARA series are suitable for internal or external circulation and have a choice of eight different controllers that are suitable for various different applications.
  • All thermostats feature an intuitive, user interface with bright display to view important readings
  • Thermostat can be turned around and positioned on all four sides on the bath for easy viewing
  • Each system comes with a quick-start guide for simple setup and operation
  • Save up to 80% of energy costs and thousands of kilowatt hours during the product lifetime
Certifications: Units areMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable