Sample vessel
Supplier: Hach
LZQ035
HACHLZQ035EA 0 GBP
HACHLZQ035
Sample vessel
Sample Containers General Purpose Containers
Sample vessel made from PS. For aqueous solutions, acids and bases.
For use with autosampler LPV122/LZQ031.