Rectangular Cuvettes with Cap
Supplier: Hach
LZM381
HACHLZM381EA 72.2 GBP
HACHLZM381
Rectangular Cuvettes with Cap
Cuvettes Spectrophotometer Cuvettes
New disposable sample cuvette with screw cap for easy and safe colour measurements. The cuvette is specially designed for the measurement of volatile, toxic or carcinogenic substances. The easily sealable rectangular sample cuvette with screw cap meets the safety requirements of the chemical and pharmazeutical industry.
  • Secure and reliable measurement results because of high beam path lengths
  • Safe handling since there the is no spillage risk due to a closed sample cuvette
  • Protection against outgassing or premature evaporation of the sample liquid