Calcium Standard Solution, 1000 mg/L as Ca (NIST), 500 ml, Hach
Supplier: Hach
2240349
HACH2240349EA 71.7 GBP
HACH2240349
Calcium Standard Solution, 1000 mg/L as Ca (NIST), 500 ml, Hach
Standards Cationic Standards Calcium Standards
For accuracy checks and instrument calibrations in calcium determinations.