Titrator, digital
Supplier: Hach
2271900 2063900 2063300 1690002
HACH2271900EA 151 GBP
HACH2271900 HACH2063900 HACH2063300 HACH1690002
Titrator, digital
Titrators
Portable and robust digital Titrator for fast and precise titrations. High resolution (125 µl/digit) and high accuracy (±1%). Applications for more than 40 parameters with interchangeable cartridges.
  • Digital Titrator methods use a high-precision dispensation device and cartridges of concentrated titrant to allow analyses in the lab, the plant or the field
  • Interchangeable cartridges allow you to change titrants in seconds; no cleaning or rinsing burettes
  • Versatile: More than 50 parameter/range combinations are available
  • Concentrated titrants allow 40 to 100
More Product Information